Order Entry
Austria
ContactUsLinkComponent
 
Beryllium standard solution, 10 mg/L Be in 5% nitric acid (from BeO.(Be(OAc)₂)₃), standard for ICP, Specpure®
Catalog # 45248.AE
CAS Number:  
Beryllium standard solution, 10 mg/L Be in 5% nitric acid (from BeO.(Be(OAc)₂)₃), standard for ICP, Specpure®
Catalog # 45248.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00134032
  • Element:
    Beryllium
  • Symbol:
    Be

Specifications

About this item

Molecular Formula: Be₄O(C₂H₃O₂)6 in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134032

Certifications: Traceable to NIST