J.T.Baker®, Antimony Standard Solution, 1000 mg/L Sb in dil. Nitric Acid (from Antimony (III) potassium oxitartrate trihydrate) Standard for AAS
Catalog # 6968.0100
Supplier: Avantor
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- Element:Antimony
- Symbol:Sb
Specifications
About this item
Certifications: Traceable to NIST