Cadmium standard solution, 100 mg/L Cd in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS08N001X1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:100 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Element:Cadmium
- Symbol:Cd