Order Entry
Austria
ContactUsLinkComponent
Lead standard solution, 1000 mg/L Pb in 0,1% nitric acid (from Pb(NO₃)₂) TraceCERT® standard for ion chromatography, Supelco®
Lead standard solution, 1000 mg/L Pb in 0,1% nitric acid (from Pb(NO₃)₂) TraceCERT® standard for ion chromatography, Supelco®
Catalog # 51777-100ML
Supplier:  Merck
CAS Number:  
undefined
Lead standard solution, 1000 mg/L Pb in 0,1% nitric acid (from Pb(NO₃)₂) TraceCERT® standard for ion chromatography, Supelco®
Catalog # 51777-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,1% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item