Order Entry
Austria
ContactUsLinkComponent
Aluminium standard solution, 10 mg/L Al in dil. nitric acid
Aluminium standard solution, 10 mg/L Al in dil. nitric acid
Catalog # CMPLF562.10.2N.L5
CAS Number:  
Aluminium standard solution, 10 mg/L Al in dil. nitric acid
Catalog # CMPLF562.10.2N.L5
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item