Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Acetic acid
Supelco® Organic Reference Standard, Acetic acid
Catalog # 71251-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Acetic acid
Catalog # 71251-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    64-19-7
  • Description:
    Acetic acid
  • Storage conditions:
    RT
  • Hazard class description:
    H226 - H314
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00036152
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Glacial acetic acid
  • Melting point:
    16.2 °C (lit.)
  • Formula:
    CH₃CO₂H
  • Molecular weight:
    60,05
  • Density:
    1,049 g/ml at 25 °C (lit.)
  • Flash point:
    39 °C
  • Boiling point:
    117 - 118 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS05
Caution: P210 - P233 - P240 - P280 - P303 + P361 + P353 - P305 + P351 + P338