Order Entry
Austria
ContactUsLinkComponent
 
Formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
 
Formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™

 

  • Assay of formic acid:
    0.093 - 0.107 %
  • pH (20 °C):
    2.6 - 2.8
  • Calcium (Ca):
    Max. 0.5 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Sodium (Na):
    Max. 2 ppm
  • Transmittance at 210 nm:
    Min. 5 %
  • Transmittance at 230 nm:
    Min. 45 %
  • Transmittance at 254 nm:
    Min. 98 %
  • HPLC Gradient at 210 nm:
    Max. 50 mAU
  • HPLC Gradient at 254 nm:
    Max. 10 mAU
  • LC-MS application:
    complying

 

Frequently Bought Together