Order Entry
Austria
ContactUsLinkComponent
Potassium bromate concentrate for dilution to 1L, 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Health
Irritant
Potassium bromate concentrate for dilution to 1L, 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Catalog # 38080-1EA
CAS Number:  
Health
Irritant
Potassium bromate concentrate for dilution to 1L, 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Catalog # 38080-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :167 g/mol
Density:1.024 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-829-8
ADR:5.1,II
 :GHS08
 :GHS07
CAS Number:7758-01-2
Formula:KBrO₃
MDL Number:MFCD00011359