Sodium hydrogen carbonate 75 g/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:205-633-8
Frequently Bought Together