Beryllium standard solution, 1,000 mg/L Be in dil. nitric acid (from BeO.(Be(OAc)₂)₃) standard for AAS and ICP
Catalog # PERCN9300172
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:125 mL
- Element:Beryllium
- Symbol:Be