Magnesium chloride 510 g/l in aqueous solution
Catalog # HACH1476253
Supplier: Hach
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
:
:95.21 g/mol
Boiling Pt:1412
Melting Pt:715
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106
Merck Index:13,05684