

Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:77 °C
Density:0.886 g/cm3
Flash Pt:22 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance of the substance:complying
- Identity (IR):complying
- Assay:69.2 - 71.5 Vol.%
- Density (D 20/20):0.883 - 0.889
- Non-volatile matter:Max. 25 mg/l
- Acidity or alkalinity:complying
- Acetaldehyde & Acetale (GC):Max. 8 Vol.ppm
- Benzene (GC):Max. 2 Vol.ppm
- Methanol (GC):Max. 150 Vol.ppm
- Impurities (GC) total:Max. 250 Vol.ppm
- Volatile impurities (GC):complying
Specification Test Results
Frequently Bought Together