Order Entry
Austria
ContactUsLinkComponent
D-(+)-Glucose monohydrate 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
D-(+)-Glucose monohydrate 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(+)-Glucose monohydrate 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C: Water (H₂O):
    7.5 - 9.5 %
  • USP - Assay (Anhydrous basis):
    97.5 - 102.0 %
  • USP - Related substances: Maltose and isomaltose:
    ≤0.4 %
  • USP - Related substances: Maltotriose:
    ≤0.2 %
  • USP - Related substances: Fructose:
    ≤0.15 %
  • USP - Related substances: Unspecified impurities, each:
    ≤0.10 %
  • USP - Related substances: Total Impurities:
    ≤0.5 %
  • USP - Color and Clarity of Solution:
    Passes Test
  • USP - Soluble Starch, Sulfites:
    ≤15 ppm
  • USP - Conductivity (uS cm-1):
    ≤20 µS/cm
  • USP - Dextrin:
    Passes Test
  • USP - Endotoxin Concentration (EU/g):
    ≤2.5
  • EP/BP - Identification A (Specific Rotation (+)):
    52.5 - 53.3 Degree
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification E:
    Passes Test
  • EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance):
    97.5 - 102.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Conductivity, uS cm-1:
    ≤20
  • EP/BP - Related Substances: sum of impurities A and B:
    ≤0.4 %
  • EP/BP - Related Substances: impurity C:
    ≤0.2 %
  • EP/BP - Related Substances: impurity D:
    ≤0.15 %
  • EP/BP - Related Substances: unspecified impurities, each:
    ≤0.10 %
  • EP/BP - Related Substances: total impurities:
    ≤0.5 %
  • EP/BP - Dextrins:
    Passes Test
  • EP/BP - Soluble starch, sulfite:
    ≤15 ppm
  • EP/BP - Water (H₂O):
    7.0 - 9.5 %
  • JP - Identification 1:
    Passes Test
  • JP - Identification 2:
    Passes Test
  • JP - Water (H2O):
    7.5 - 9.5 %
  • JP - Assay (Anhydrous basis):
    97.5 - 102.0 %
  • JP - Maltose and isomaltose:
    ≤0.4 %
  • JP - Maltotriose:
    ≤0.2 %
  • JP - Fructose:
    ≤0.15 %
  • JP - Unspecified Impurities, each:
    ≤0.10 %
  • JP - Total Impurities:
    ≤0.5 %
  • JP - Color and Clarity of Solution:
    Passes Test
  • JP - Soluble Starch, Sulfites:
    ≤15 ppm
  • JP - Conductivity (uS cm-1):
    ≤20 µS/cm
  • JP - Dextrin:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤4 ppm

Specification Test Results

Related Information

Frequently Bought Together