Lead standard solution, 1000 mg/L Pb in dil. nitric acid (from Pb) standard for AAS and ICP
Supplier: PerkinElmer
Certificates
Lead standard solution, 1000 mg/L Pb in dil. nitric acid (from Pb) standard for AAS and ICP
Supplier: PerkinElmer
About this item
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals