Order Entry
Austria
ContactUsLinkComponent
 
Irritant
di-Ammonium oxalate monohydrate ≥96%, high purity
Catalog # E742-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
di-Ammonium oxalate monohydrate ≥96%, high purity
Catalog # E742-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Purity:
    >96%
  • Residue on Ignition:
    <0.05%

Specification Test Results

About this item

 :
 :142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:∼1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-202-3
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:6009-70-7
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694