Cadmium standard solution, 1,000 mg/L Cd in 0.5 M nitric acid standard for AAS
Supplier: Thermo Fisher Scientific
Certificates
Cadmium standard solution, 1,000 mg/L Cd in 0.5 M nitric acid standard for AAS
Supplier: Thermo Fisher Scientific
About this item
Frequently Bought Together
BUFFER PH 10 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
UV/Visible standards, Supelco®
Supplier: Merck
VWR® Conductivity standard, CRM, 5000 μS/cm
Supplier: VWR Chemicals
Ammonium CRMs, standard solutions, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
4'-Ethoxyacetanilide, Supelco®
Supplier: Merck
DL-α-Tocopherol acetate, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Turbidity standards, Stablcal®
Supplier: Hach
Geraniol
Supplier: EHRENSTORFER
SCANDIUM STANDARD FOR ICP 1 * 100 mL
Supplier: Merck