Orders LinkContactUsLinkComponent
Half area microplates, 96-well
Half area microplates, 96-well
   Sustainable Options Available
Supplier:   Greiner Bio-One
Half area microplates, 96-well
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
Half area microplates, 96-well
7 Options Available Below
Product Details & Documents
PS, without lid. 96-well half area microplates offer an alternative to 384-well or 1536-well microplates when low sample volume is an important criterion but automation is not necessarily appropriate. They can be pipetted manually while at the same time allow a reduction in sample volume of up to 50%.
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardised pathlength, eliminating the need for pathlength determinations. White plates are ideal for luminescence assays, while black plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
Specifications for Product Family
Specifications
  • Catalog No:
  • GREI675161_40
  • 738-0145
  • GREI675074_40
  • 392-0287
  • GREI675077_40
  • 392-0288
  • 392-0290
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
  • Colour
    Sterile
    Availability
    Price
  • GREI675161_40
    Half area, solid bottom, non treated surface
    Half area, solid bottom, non treated surface
    Clear
    +
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 158,00 €
    158,00 €
    1 * 40 ST
       158,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI675161_40
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0145
    Half area, solid bottom, non treated surface
    Half area, solid bottom, non treated surface
    Clear
    -
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 110,00 €
    110,00 €
    1 * 40 ST
       110,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0145
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI675074_40
    Half area, solid bottom, high binding surface
    Half area, solid bottom, high binding surface
    White
    +
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 151,00 €
    151,00 €
    1 * 40 ST
       151,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI675074_40
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 392-0287
    Half area, solid bottom, medium binding surface
    Half area, solid bottom, medium binding surface
    White
    -
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 137,00 €
    137,00 €
    1 * 40 ST
       137,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    392-0287
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI675077_40
    Half area, solid bottom, high binding surface
    Half area, solid bottom, high binding surface
    Black
    +
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 226,00 €
    226,00 €
    1 * 40 ST
       226,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI675077_40
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 392-0288
    Half area, solid bottom, medium binding surface
    Half area, solid bottom, medium binding surface
    Black
    -
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 136,00 €
    136,00 €
    1 * 40 ST
       136,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    392-0288
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 392-0290
    Half area, µClear® bottom, medium binding surface
    Half area, µClear® bottom, medium binding surface
    Black/clear
    -
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 207,00 €
    207,00 €
    1 * 40 ST
       207,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    392-0290
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact