Order Entry
Austria
Orders LinkContactUsLinkComponent
Multi-function stopwatch

Multi-function stopwatch

Stopwatch, 60 min, Yellow/blue, 87×21×78 mm
Catalog # HEATHS24645
Supplier:   HEATHROW
Multi-function stopwatch
Catalog # HEATHS24645
Supplier:  HEATHROW
Multi-function stopwatch
Multi-function stopwatch

Stopwatch, 60 min, Yellow/blue, 87×21×78 mm

Price
73,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Timing capacity: 60 minutes in ¹/₁₀₀ second increments.

  • Lightweight, dependable, accurate and easy to use
  • Large ¹/₄" display
  • Functions: split, hourly chime, calendar, alarm

Delivery information: Supplied with neck cord and one LR44 (1,5 V) battery.

Specifications
  • Colour:
    Yellow/blue
  • Description:
    Stopwatch, 60 min
  • W×D×H:
    87×21×78 mm