Orders LinkContactUsLinkComponent
Stopcocks with GL thread, EX
Stopcocks with GL thread, EX
Supplier:   Bohlender
Stopcocks with GL thread, EX
Supplier:  Bohlender
Certificates
Stopcocks with GL thread, EX
Stopcocks with GL thread, EX
9 Options Available Below
Product Details & Documents
Stopcocks with GL thread for distributing liquids or gases, quick and easy disconnection of flow.
  • Cylindrical stopcock plug made of conductive PTFE-EX for good tightness
  • Stop valve with mark of flow direction
  • Grip made of red PP
  • Suitable for pressure up to maximum 6 bar and suitable for vacuum
  • Temperature resistance upto 110 °C
  • Chemical resistant
Two-way stopcock made of conductive PTFE-EX with straight bore and two connections with GL thread or three-way stopcock with L-shaped or T-shaped bore and three connections with GL thread.
Product Family Options
2-way stopcocks, straight
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 8 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 6 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • 4 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
3-way stopcocks, L-shaped bore
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 4 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
  • 6 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 177,00 €
    177,00 €
    1 * 1 ST
       177,00 €
  • 6 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 265,00 €
    265,00 €
    1 * 1 ST
       265,00 €
3-way stopcocks, T-shaped bore
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 4 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 6 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • 4 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 188,00 €
    188,00 €
    1 * 1 ST
       188,00 €