Orders LinkContactUsLinkComponent
Microscope slide for LCP
Microscope slide for LCP
LCP sandwich set, 128×85 (slide) mm
112×77 (cover slip) mm
Catalog # MARI0890003
Supplier:   MARIENFELD
Microscope slide for LCP
Catalog # MARI0890003
Supplier:  MARIENFELD
Microscope slide for LCP
Microscope slide for LCP
LCP sandwich set, 128×85 (slide) mm
112×77 (cover slip) mm
Price
464,05 €
Log in for availability
1 * 20 ST
Product Details & Documents
The lipidic cubic phase (LCP) sandwich set consists of a base slide and a cover slip and is optimized for the crystallographic study of membrane proteins.
  • Surface with hydrophobic glass
  • Optimised for brightfield, UV and fluorescent microscopy
  • Can be used in G protein–coupled receptors (GPCR) structural biology
Specifications
  • Description:
    LCP sandwich set
  • L×W:
    128×85 (slide) mm
    112×77 (cover slip) mm