Orders LinkContactUsLinkComponent
Trypsinizing flasks, WHEATON®
Trypsinizing flasks, WHEATON®
Supplier:   DWK Life Sciences
Trypsinizing flasks, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Trypsinizing flasks, WHEATON®
Trypsinizing flasks, WHEATON®
8 Options Available Below
Product Details & Documents
Borosilicate glass, clear.
  • With pour out and caps
  • Fluted design improves mixing
  • Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Product Family Options
Graduated
Product Information
  • Capacity
    Neck
    Availability
    Price
  • WHEA355803
    Top screw cap 33-430
    Top screw cap 33-430
    75 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 232,00 €
    232,00 €
    1 * 1 ST
       232,00 €
  • WHEA355804
    Top screw cap 38-430
    Top screw cap 38-430
    150 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 238,00 €
    238,00 €
    1 * 1 ST
       238,00 €
  • WHEA355807
    Top screw cap 51-400
    Top screw cap 51-400
    700 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 317,00 €
    317,00 €
    1 * 1 ST
       317,00 €
Non graduated
Product Information
  • Capacity
    Neck
    Availability
    Price
  • WHEA355754
    Top screw cap 38-430
    Top screw cap 38-430
    150 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €
  • 734-3116
    Top screw cap 33-430
    Top screw cap 33-430
    75 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 190,00 €
    190,00 €
    1 * 1 ST
       190,00 €
  • 392-0369
    Top screw cap 24-430
    Top screw cap 24-430
    35 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €
  • WHEA355755
    Top screw cap 38-430
    Top screw cap 38-430
    300 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • WHEA355757
    Top screw cap 51-400
    Top screw cap 51-400
    700 ml
    Screw cap 20-415
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €