Counting chambers
Supplier: Kova International
Jump to:
About this item
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
- High optical quality through the composition acrylate
- Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
- Capillary action for reproducible precision
- Homogeneous suspension of sample sediments: Retained volume of 1 µl
- For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl
5 Options Available Below
Product Details & Documents
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
- High optical quality through the composition acrylate
- Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
- Capillary action for reproducible precision
- Homogeneous suspension of sample sediments: Retained volume of 1 µl
- For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl
Documents
Product Family Options
Product Information
- TypeW×DAvailabilityPrice
- 630-1506Slides with ten chambers and patented counting grids (table included)Slides with ten chambers and patented counting grids (table included)with grid3×3 mmLog in for availability1 * 100 ST231,00 €
- 630-1539Slides with ten chambers and large viewing areaSlides with ten chambers and large viewing areawithout grid3×3 mmLog in for availability1 * 100 ST195,00 €
- 630-1537Slides with four chambers and counting gridsSlides with four chambers and counting gridswith grid3×3 mmLog in for availability1 * 100 ST162,00 €
- 630-1538Slides with four chambers
- 630-2048Slides with ten chambers and a large viewing areaSlides with ten chambers and a large viewing areawithout grid3×3 mmLog in for availability1 * 1 SET112,00 €