Orders LinkContactUsLinkComponent
Percoll™
Percoll™
Supplier:   Cytiva
Percoll™
Supplier:  Cytiva
Certificates
Percoll™
3 Options Available Below
Product Details & Documents
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For Research Use Only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0,25 M sucrose, and for formation of gradients either by the use of convenient gradient mixers or by high speed centrifugation. Colloidal solution of silica coated with polyvinylpyrrolidone (PVP). Liquid/suspension. Colorless to slightly yellow. Certificate of Analysis is available.
Caution: Percoll is for Research use only.
Product Family Options
Product Information
  • Availability
    Price
  • Percoll™
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 222,00 €
    222,00 €
    1 * 250 mL
       222,00 €
  • Percoll™
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 530,00 €
    530,00 €
    1 * 1 L
       530,00 €
  • 17-0891-09
    Percoll™, 6×1 l
    Percoll™, 6×1 l
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 3 100,00 €
    3 100,00 €
    1 * 6 ST
       3 100,00 €