ContactUsLinkComponent
illustra™ TempliPhi™100/500 Amplification Kits
illustra™ TempliPhi™100/500 Amplification Kits
Supplier:  Cytiva
Certificates
undefined
illustra™ TempliPhi™100/500 Amplification Kits
Supplier:  Cytiva
Specifications
  • Storage conditions:
    2 months or less at -20°C (for longer, store at -70°C). Thaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification.