Order Entry
Austria
ContactUsLinkComponent
 
illustra™ MicroSpin™ S-400 HR columns
  27-5140-01
 :  Cytiva
 :  
undefined
illustra™ MicroSpin™ S-400 HR columns
  27-5140-01
 :  Cytiva
 :  

 

  • Pk:
    50
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Illustra™ MicroSpin™ S-400 HR columns
  • Description:
    Illustra™ MicroSpin™ S-400 HR columns

 

 

illustra™ MicroSpin™ S-400 HR columns designed for rapid purification of PCR products (>200 bp) from unincorporated primers (<32-mers) and nucleotides using spin-column chromatography.

  • Accommodates 25 to 50 μl for post-PCR cleanup prior to cloning or a second amplification reaction or 51 to 100 μl for all other applications
  • Prepacked for convenience with Sephacryl™ S-400 HR pre-equilibriated in TE buffer
  • Ready to use, requiring less than 4 minutes from sample application to collection of purified product
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays