ContactUsLinkComponent
Syringes for HPLC autosamplers, Spark Holland instruments
Syringes for HPLC autosamplers, Spark Holland instruments
Catalog # 548-1646
undefined
Syringes for HPLC autosamplers, Spark Holland instruments
Catalog # 548-1646
Specifications
  • For:
    Spark Holland Promis LC autosampler
  • Type:
    250D-SPARK-GT
  • Volume:
    250 µl
  • Description:
    Gas-tight, with PTFE-tipped plunger, 1/4-28 UNF front thread
  • Seq:
    01
  • Pk:
    1