Fork wrenches, BOLA
Supplier: Bohlender
Jump to:
About this item
These glass fibre reinforced PA fork wrenches can be used for tightening or opening screw joints also at high working temperatures.
- Good chemical resistance
- Temperature resistant from −10 to +140 °C
- Low weight reduces risk of injury or damage
2 Options Available Below
Product Details & Documents
These glass fibre reinforced PA fork wrenches can be used for tightening or opening screw joints also at high working temperatures.
- Good chemical resistance
- Temperature resistant from −10 to +140 °C
- Low weight reduces risk of injury or damage
Product Family Options
Product Information
- MaterialAvailabilityPrice
- BOHLD647-08Fork wrench for thread GL 14/GL 18/GL 25
- BOHLD647-24Fork wrench for thread GL 32/GL 45