Order Entry
Austria
ContactUsLinkComponent
Visual colour kits, sulphite, MQuant®, Supelco®
Visual colour kits, sulphite, MQuant®, Supelco®
Catalog # 1.11148.0001
Supplier:  Merck
CAS Number:  
Visual colour kits, sulphite, MQuant®, Supelco®
Catalog # 1.11148.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Chemical method:
    Iodate/starch
  • Pk:
    200 Tests
  • Description:
    Sulphite test
  • M-Clarity quality level:
    MQ100
  • Type:
    MTC
  • Range:
    0,5 - 50 mg/l Na₂SO₃, 0,3-32 mg/l SO₃

Specifications

About this item

MQuant® titrimetric and colorimetric methods.

  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.