Multiple distributors for bottles, EX, BOLA
About this item
Black screw cap, made of conductive PPS-EX, for GL 45 threaded bottles and distributor body made of conductive PTFE-EX.
- Very good chemical resistance
- Temperature resistant from −20 to +200 °C
- Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of 8,5 mm can be inserted through the necks.
2 Options Available Below
Product Details & Documents
Black screw cap, made of conductive PPS-EX, for GL 45 threaded bottles and distributor body made of conductive PTFE-EX.
- Very good chemical resistance
- Temperature resistant from −20 to +200 °C
- Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of 8,5 mm can be inserted through the necks.
Product Family Options
Product Information