Orders LinkContactUsLinkComponent
Supelco® Reflectoquant® Test Strips, Peroxide
Supelco® Reflectoquant® Test Strips, Peroxide
Supplier:  Merck
Certificates
Supelco® Reflectoquant® Test Strips, Peroxide
3 Options Available Below
Product Details & Documents
Test strips for the reflectometers RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
This test is suited for the determination of inorganic peroxides in aqueous solutions. Furthermore, it can be used in disinfecting and rinsing solutions (e.g., in food technology and laundries) and in swimming pool water.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.16974.0001
  • 1.17968.0001
  • 1.16731.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    Type
    Pk
    Availability
    Price
  • Peroxide test
    0,2 - 20,0 mg/L H₂O₂
    Enzymatic reaction
    RQ
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 67,80 €
    67,80 €
    1 * 50 Tests
       67,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.16974.0001
    M-Clarity quality level
    MQ100
  • Peroxide test
    20 - 100 mg/L H₂O₂
    Enzymatic reaction
    RQ
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 136,00 €
    136,00 €
    1 * 50 Tests
       136,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.17968.0001
    M-Clarity quality level
  • 1.16731.0001
    Peroxide test
    Peroxide test
    100 - 1000 mg/L H₂O₂
    Enzymatic reaction
    RQ
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 84,20 €
    84,20 €
    1 * 50 Tests
       84,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.16731.0001
    M-Clarity quality level
    MQ100
Complementary Products
Product Information
  • Catalog #
    Description
    Model
    Availability
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log in for availability
    1 * 1 ST - 1 420,00 €