ContactUsLinkComponent
Reflectoquant®10 plus Test kits, Supelco®
Reflectoquant®10 plus Test kits, Supelco®
Catalog # 1.17945.0001
Supplier:  Merck
CAS Number:  
Reflectoquant®10 plus Test kits, Supelco®
Catalog # 1.17945.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Kalignost
  • Pk:
    100 Tests
  • Description:
    Reflectoquant® 10 plus Potassium test
  • M-Clarity quality level:
    MQ100
  • Range:
    1,0 - 25,0 mg/l K
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly