ContactUsLinkComponent
Test strips, total sugar (glucose and fructose), Reflectoquant®, Supelco®
Catalog # 1.16136.0001
Supplier:  Merck
CAS Number:  
Test strips, total sugar (glucose and fructose), Reflectoquant®, Supelco®
Catalog # 1.16136.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Enzymatic reaction
  • Pk:
    50 Tests
  • Description:
    Total sugar test (glucose and fructose)
  • M-Clarity quality level:
    MQ100
  • Range:
    65 - 650 mg/l
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly