Orders LinkContactUsLinkComponent
Test strips, phosphate, MQuant®, Supelco®
Test strips, phosphate, MQuant®, Supelco®
Phosphate test, 10-25-50-100-250-500 mg/l PO₄, Phosphormolybdenum blue, MQ, 100 Tests
Catalog # 1.10428.0001
Supplier:   Merck
Test strips, phosphate, MQuant®, Supelco®
Catalog # 1.10428.0001
Supplier:  Merck
CAS Number:  
Test strips, phosphate, MQuant®, Supelco®
Test strips, phosphate, MQuant®, Supelco®
Phosphate test, 10-25-50-100-250-500 mg/l PO₄, Phosphormolybdenum blue, MQ, 100 Tests
CAS Number:  
Price
78,50 €
Log in for availability
1 * 100 Tests
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: Just dip, add reagent, wait and measure visually against colour scale
  • Immediate results within a few minutes enables fast decisions
  • Portable: Can be used inside or outside the laboratory
Documents
Specifications
Specifications
  • Chemical method:
    Phosphormolybdenum blue
  • Pk:
    100 Tests
  • Description:
    Phosphate test
  • Type:
    MQ
  • Range:
    10-25-50-100-250-500 mg/l PO₄
  • Cat. No.:
    1.10428.0001
  • M-Clarity quality level:
    MQ100