ContactUsLinkComponent
Test strips and reagents for semi-quantitative determinations of phosphate in water analyses, QUANTOFIX®
Test strips and reagents for semi-quantitative determinations of phosphate in water analyses, QUANTOFIX®
Catalog # 705-0320
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips and reagents for semi-quantitative determinations of phosphate in water analyses, QUANTOFIX®
Catalog # 705-0320
Supplier:  MACHEREY-NAGEL
CAS Number:  
Specifications
  • Chemical method:
    Molybdenum blue
  • Pk:
    100 Tests
  • Description:
    Phosphate
  • Seq:
    35
  • Environmentally Preferable:
  • Range:
    0 - 100 mg/l PO₄3-