Orders LinkContactUsLinkComponent
Test strips, nitrite, MQuant®, Supelco®
Test strips, nitrite, MQuant®, Supelco®
Test strips, nitrite, MQuant®, Supelco®
Anbieter:  Merck
Certificates
Test strips, nitrite, MQuant®, Supelco®
4 Options Available Below
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.10022.0001
  • 1.10057.0001
  • 1.10007.0002
  • 1.10007.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Description
    Range
    Chemical method
    Type
    Pk
    Verfügbarkeit
    Preis
  • Nitrite test
    0,1-0,3-0,6-1-2-3 g/l NO₂
    Griess' reaction
    MQ
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 45,70 €
    45,70 €
    1 * 100 Tests
       45,70 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    1.10022.0001
    M-Clarity quality level
    MQ100
  • Nitrite test
    0-0,5-1-2-5-10 mg/l NO₂
    Griess' reaction
    MQ
    75 Tests
    Log in for availability
    Log in for availability
    1 * 75 Tests
    1 * 75 Tests - 59,00 €
    59,00 €
    1 * 75 Tests
       59,00 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    1.10057.0001
    M-Clarity quality level
    MQ100
  • Nitrite test
    2-5-10-20-40-80 mg/l NO₂
    Griess' reaction
    MQ
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests
    Specifications:
    Mehr
    Specifications
    Cat. No.
    1.10007.0002
    M-Clarity quality level
    MQ100
  • Nitrite test
    2-5-10-20-40-80 mg/l NO₂
    Griess' reaction
    MQ
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 69,00 €
    69,00 €
    1 * 100 Tests
       69,00 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    1.10007.0001
    M-Clarity quality level
    MQ100