Test Strips, Oil
About this item
This test paper allows the rapid and reliable detection of oil contamination of water and soil.
- Colour changes: Blue to dark blue
For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample.
3 Options Available Below
Product Details & Documents
This test paper allows the rapid and reliable detection of oil contamination of water and soil.
- Colour changes: Blue to dark blue
For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample.
Documents
Certificates
Search by Lot NumberDip and Read Tests, Rapid Tests.pdf
Additional Product Details
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 705-0312
- 705-0322
- MANA91338
- Method:
- Nitrite - Griess reaction (white → red-violet)
- Nitrite - Griess reaction (yellow → red)
- Nitrite - Griess reaction (white → red-violet);pH - mixed indicator (yellow-orange → violet-red)
- Parameters:
- All reagents included
- All reagents included
- All reagents included
- Compliance:
- No hazardous material
- No hazardous material
- No hazardous material
- Contents:
- 100 test strips in a tube, instruction leaflet
- 100 test strips in a tube, instruction leaflet
- 100 test strips in a tube, instruction leaflet
- Instrument:
- QUANTOFIX Relax
- QUANTOFIX Relax
- No
- Platform:
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Shelf life (months):
- 30 (from production)
- 30 (from production)
- 30 (from production)
- Weight:
- 44,3 g
- 40,3 g
- 35 g
Specifications
Product Family Options
Product Information
- RangeChemical methodDimensionsTests per kitPkAvailabilityPrice
- 705-0312QUANTOFIX® nitriteQUANTOFIX® nitrite0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 0,5–80 - mg/L - NO₂⁻Azo reaction with an aromatic amine40×40×120 mm (packaging)100 Tests100 TestsLog in for availability1 * 100 Tests34,50 €Specifications:
More
Specifications
Cat. No.705-0312MethodNitrite - Griess reaction (white → red-violet)ParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tube, instruction leafletInstrumentQUANTOFIX RelaxPlatformSemi-quantitative test stripsShelf life (months)30 (from production)Weight44,3 g - 705-0322QUANTOFIX® nitrite 3000QUANTOFIX® nitrite 30000 • 0,1 • 0,3 • 0,6 • 1 • 2 • 3 - g/L - NO₂⁻; 0,1–3,0 - g/L - NO₂⁻Azo reaction with an aromatic amine40×40×120 mm (packaging)100 Tests100 TestsLog in for availability1 * 100 Tests34,50 €Specifications:
More
Specifications
Cat. No.705-0322MethodNitrite - Griess reaction (yellow → red)ParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tube, instruction leafletInstrumentQUANTOFIX RelaxPlatformSemi-quantitative test stripsShelf life (months)30 (from production)Weight40,3 g - MANA91338QUANTOFIX® nitrite/pHQUANTOFIX® nitrite/pH0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 6,0 • 6,4 • 6,7 • 7,0 • 7,3 • 7,6 • 7,9 • 8,2 • 8,4 • 8,6 • 8,8 • 9,0 • 9,3 • 9,6Nitrite: Azo reaction with an aromatic amine; pH: pH indicator43×123×40 mm (packaging)100 Tests100 TestsLog in for availability1 * 100 Tests42,90 €Specifications:
More
Specifications
Cat. No.MANA91338MethodNitrite - Griess reaction (white → red-violet);pH - mixed indicator (yellow-orange → violet-red)ParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tube, instruction leafletInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)30 (from production)Weight35 g
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



