About this item
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: just dip, wait and measure visually against colour scale
- Safe: no auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: can be used inside or outside the laboratory
3 Options Available Below
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: just dip, wait and measure visually against colour scale
- Safe: no auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: can be used inside or outside the laboratory
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1.10020.0001
- 1.10020.0002
- 1.10092.0021
- M-Clarity quality level:
- MQ100
- MQ100
- MQ100
Specifications
Product Family Options
Product Information
- RangeChemical methodTypePkAvailabilityPrice
- 1.10020.0001Nitrate testNitrate test10-25-50-100-250-500 mg/l NO₃Griess' reaction (modified)MQ100 TestsLog in for availability1 * 100 Tests60,00 €Specifications:
More
Specifications
Cat. No.1.10020.0001M-Clarity quality levelMQ100 - 1.10020.0002Nitrate testNitrate test10-25-50-100-250-500 mg/l NO₃Griess' reaction (modified)MQ25 TestsLog in for availability1 * 25 Tests32,00 €Specifications:
More
Specifications
Cat. No.1.10020.0002M-Clarity quality levelMQ100 - 1.10092.0021Nitrate test ( individually sealed)Nitrate test ( individually sealed)10-25-50-100-250-500 mg/l NO₃Griess' reaction (modified)MQ1.000 TestsLog in for availability1 * 1.000 Tests455,00 €Specifications:
More
Specifications
Cat. No.1.10092.0021M-Clarity quality levelMQ100