Order Entry
Austria
Orders LinkContactUsLinkComponent
 

QUANTOFIX® Test Strips, Arsenic

QUANTOFIX® Test Strips, Arsenic
Supplier:  MACHEREY-NAGEL
Certificates
QUANTOFIX® Test Strips, Arsenic

2 Options Available Below

Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

The test strips allow the quick and easy determination of arsenic in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 20 to 30 minutes one gets reliable results.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 705-0316
  • MANA91345.2
  • Method:
  • Ammonium - Nessler (bright yellow → orange)
  • Arsenic - Modified Gutzeit test (white → yellow-brown)
  • Parameters:
  • Requires additional reagent
  • Requires additional reagent
  • Compliance:
  • Hazardous material
  • Hazardous material
  • Contents:
  • 100 test strips in a tube, additional regaents, instruction leaflet in a box
  • 100 test strips in a tube, additional regaents, instruction leaflet in a box
  • Instrument:
  • QUANTOFIX Relax
  • No
  • Platform:
  • Semi-quantitative test strips
  • Semi-quantitative test strips
  • Shelf life (months):
  • 30 (from production)
  • 30 (from production)
  • Weight:
  • 230 g
  • 558 g

Specifications

Product Family Options

Product Information

  • Range
    Chemical method
    Dimensions
    Tests per kit
    Pk
    Availability
    Price
  • QUANTOFIX® ammonium
    0 • 10 • 25 • 50 • 100 • 200 • 400 - mg/L - NH₄⁺; 10–350 - mg/L - NH₄⁺
    Color reaction based on Neßler’s reagent
    170×125×48 mm (packaging)
    100 Tests
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 57,50 €
    57,50 €
    1 * 100 Tests
       57,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    705-0316
    Method
    Ammonium - Nessler (bright yellow → orange)
    Parameters
    Requires additional reagent
    Compliance
    Hazardous material
    Contents
    100 test strips in a tube, additional regaents, instruction leaflet in a box
    Instrument
    QUANTOFIX Relax
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    230 g
  • MANA91345.2
    QUANTOFIX EZ Arsenic Sensitive PP
    QUANTOFIX EZ Arsenic Sensitive PP
    0 • 0.005 • 0.01 • 0.025 • 0.05 • 0.1 • 0.25 • 0.5 - mg/L - As³⁺ ⁵⁺
    200×180×135 mm (package)
    100 test(s)
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 211,00 €
    211,00 €
    1 * 1 KIT
       211,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    MANA91345.2
    Method
    Arsenic - Modified Gutzeit test (white → yellow-brown)
    Parameters
    Requires additional reagent
    Compliance
    Hazardous material
    Contents
    100 test strips in a tube, additional regaents, instruction leaflet in a box
    Instrument
    No
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    558 g