Order Entry
Austria
ContactUsLinkComponent
Visual colour kits, pH, MQuant®, Supelco®
Visual colour kits, pH, MQuant®, Supelco®
  1.08027.0001
 :  Merck
 :  
Visual colour kits, pH, MQuant®, Supelco®
  1.08027.0001
 :  Merck
 :  

 

  • Chemical method:
    Mixed indicator
  • Pk:
    400 Tests
  • Description:
    pH test
  • Seq:
    02
  • M-Clarity quality level:
    MQ100
  • Type:
    MTC
  • Range:
    pH 4,5-5-5,5-6-6,5-7-7,5-8-8,5-9

 

 

Quick tests, visual colour comparison.

  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

 : Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.