High pressure syringes
Supplier: Trajan Scientific and Medical
Certificates
About this item
These high pressure syringes are designed for high pressure gas sampling.
- Rated to 34 bar
- Fitted with a gas tight on/off valve and high pressure 70 mm, 23 gauge needle
Replacement needles
Product Information
Frequently Bought Together