Order Entry
Austria
ContactUsLinkComponent
 
Blot module, miniVE™
  80-6418-96
 :  Cytiva
undefined
Blot module, miniVE™
  80-6418-96
 :  Cytiva

 

  • Description:
    Blot module, miniVE
  • Model:
    miniVE
  • Gel size W×L:
    100×105 mm
  • Pk:
    1

 

 

Perform 1-D and 2-D electrophoresis and electrotransfer in a single compact unit.

  • Complete semi-wet blotting in 45 minutes with only 300 ml buffer
  • Choose precast gels from a variety of manufacturers
  • Perform two second-dimension separations using 7 cm IPG strips in 15 hours

Cast, run, and blot mini-gels in four streamlined components: Gel module, blot module, lower buffer chamber, safety lid.

Two mini-gels or four mini-blots at once, using 100×105 mm gel cassettes for increased separation distances.