

4,4'-Bis(dimethylamino)diphenylmethane 98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:101-61-1
:
Formula:C₁₇H₂₂N₂
MDL Number:MFCD00008317
:254.38 g/mol
Melting Pt:88...89 °C (lit.)
:M-Clarity™ quality level = MQ100
Flash Pt:178 °C (closed cup)
EINECS:202-959-2
:GHS08
:GHS09
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour); White to Faint Beige to Tan:Conforms to Requirements
- Appearance (Form); Powder, Flakes, Crystals, Crystalline Powder and/or Chunks:Conforms to Requirements
- Infrared spectrum:Conforms to Structure
- Purity (Titration by HClO₄):97.5 - 102.5 %
- Purity (GC):≥97.5%
Specification Test Results
Frequently Bought Together