Order Entry
Austria
ContactUsLinkComponent
Lithium aluminum hydride 2 mol/l in THF, Sigma-Aldrich®
Lithium aluminum hydride 2 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL593702-100ML
Supplier:  Merck
CAS Number:  
Lithium aluminum hydride 2 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL593702-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :37.95 g/mol
ADR:4.3,I
 :M-Clarity™ quality level = MQ200
EINECS:240-877-9
UN:1411
CAS Number:16853-85-3
Formula:H₄AlLi
MDL Number:MFCD00011075