


Silver nitrate 2.5% for microscopy, Sigma-Aldrich®
Catalog # 85193-100ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance (Colour):Green to Very Dark Green
- Appearance (form):Powder or Crystals
- Argentometr. Titration:80 - 120%
- Thin layer chromatography:Corresponds to standard
- Assay (atomic absorpt.):Zn (theory) 8.74%, typically 3 - 9%
- Atomic absorption:Zn
- Solubility (Colour):0.01 g in 10 ml methanol
- Solubility (method):0.01 g in 10 ml methanol
- Loss on drying:≤8.0 %
- Sulphated ash:7 - 23 %
- Wavelength (1) (UV):621 - 636 nm
- Extinction (1%/1 cm) (1) (UV):≥800
- Solvent (UV):Water
Specification Test Results
About this item
:
:169.87 g/mol
EINECS:231-853-9
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414