Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 36% in aqueous solution analytical reagent
Corrosive
Sodium hydroxide 36% in aqueous solution analytical reagent
Catalog # 1.05596.5000
Supplier:  Merck
CAS Number:  
Corrosive
Sodium hydroxide 36% in aqueous solution analytical reagent
Catalog # 1.05596.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :M-Clarity™ quality level = MQ300
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548