Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 27% in aqueous solution, EMSURE® for nitrogen analysis, Supelco®
Corrosive
Sodium hydroxide 27% in aqueous solution, EMSURE® for nitrogen analysis, Supelco®
Supplier:  Merck
Certificates
Corrosive
Sodium hydroxide 27% in aqueous solution, EMSURE® for nitrogen analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric):
    ≥ 27.0 %
  • Carbonate (as Na₂CO₃):
    ≤ 1.0 %
  • Chloride (Cl):
    ≤ 0.004 %
  • Phosphate (PO₄):
    ≤ 0.0005 %
  • Silicate (SiO₂):
    ≤ 0.001 %
  • Sulfate (SO₄):
    ≤ 0.003 %
  • Total nitrogen (N):
    ≤ 0.0001 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • Al (Aluminium):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.0005 %

Specification Test Results

Related Information

Frequently Bought Together