Order Entry
Austria
ContactUsLinkComponent
Tris(2-ethylhexyl)phosphate for synthesis, Sigma-Aldrich®
Tris(2-ethylhexyl)phosphate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Tris(2-ethylhexyl)phosphate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Hill Formula:
    C₂₄H₅₁O₄P
  • Chemical formula:
    [CH₃(CH₂)₃CH(C₂H₅)CH₂O]₃PO
  • Molar Mass:
    434.63 g/mol
  • Boiling point:
    210 °C (5 hPa) decomposes
  • Density:
    0.92 g/cm³ (20 °C)
  • Flash point:
    170 °C
  • Ignition temperature:
    370 °C
  • pH value:
    7 (H₂O, 20 °C)
  • Vapor pressure:
     - 0.01 hPa (20 °C)
  • Solubility:
     - 0.001 g/l (20 °C)
  • LD 50 oral:
    LD50 Rat 37000 mg/kg
  • LD 50 dermal:
    LD50 Rabbit 20000 mg/kg
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 2 water endangering
  • R Phrase:
    R 38

Specification Test Results

Related Information

Frequently Bought Together