2-Adamantanone
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:700-58-3
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00074737
:150.22 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:43 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-847-2
Frequently Bought Together