2-Adamantanone ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 700-58-3 Molecular Formula: C10H14O Formula Weight: 150.22 Melting Point: ca 257° subl. Beilstein Reference: 1210235
Synonyms: Adamantan-2-one
- CAS Number:700-58-3
- Formula:C₁₀H₁₄O
- MDL Number:MFCD00074737
- Molecular Weight:150.22 g/mol
- Boiling Pt:228 °C (1013 hPa)
- Melting Pt:∼ 257 °C (subl.)
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:211-847-2
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 700-58-3 Molecular Formula: C10H14O Formula Weight: 150.22 Melting Point: ca 257° subl. Beilstein Reference: 1210235
Synonyms: Adamantan-2-one
- CAS Number:700-58-3
- Formula:C₁₀H₁₄O
- MDL Number:MFCD00074737
- Molecular Weight:150.22 g/mol
- Boiling Pt:228 °C (1013 hPa)
- Melting Pt:∼ 257 °C (subl.)
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:211-847-2
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



