2-Adamantanone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:700-58-3
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00074737
:150.22 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:282…286 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-847-2
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together